Sodium dihydrogen phosphate dihydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

A11316-22 A11316-36 A11316-0E
AAA11316-22EA 56.7 USD
AAA11316-22 AAA11316-36 AAA11316-0E
Sodium dihydrogen phosphate dihydrate 99%
Sodium dihydrogen phosphate dihydrate

MDL: MFCD00149209 Fieser: 1,395


Purity: 99% CAS No.: 13472-35-0 Molecular Formula: H2NaO4P.2H2O Formula Weight: 156.01 (119.98anhy) Merck Reference: 14,8660 MDL No.: MFCD00149209

Formula: NaH₂PO₄·2H₂O
MW: 156.01 g/mol
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 60
Density: 1.910
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 14,8660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR