(±)-2-Methylcaproic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-MethylhexanoicAcid, 2-Methylhexanoic acid, (±)-2-Methylhexanoic acid
A11290-14
A11290-06
AAA11290-14EA
161
USD
AAA11290-14
AAA11290-06
(±)-2-Methylcaproic acid 98%
(±)-2-Methylcaproic acid
MDL: MFCD00002674
Beilstein Registry No.: 1721227
Purity: 98%
CAS No.: 4536-23-6
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 105°(221°F)
Refractive Index: 1.4205
Odor: Rancid
Boiling Point: 209-210°
Beilstein Reference: 1721227
UN No.: 3265
MDL No.: MFCD00002674
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 209 °C (1013 hPa) Density: 0.929 g/cm³ (20 °C) Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002674 CAS Number: 4536-23-6 EINECS: 224-883-9 UN: 2920 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...