α,2-Dibromotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromobenzyl bromide

A11285-09 A11285-18
AAA11285-09EA 52.4 USD
AAA11285-09 AAA11285-18
α,2-Dibromotoluene 98%
α,2-Dibromotoluene

MDL: MFCD00000173 Beilstein Registry No.: 971015


Purity: 98% CAS No.: 3433-80-5 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Flash Point: 110°(230°F) Refractive Index: 1.6190 Melting Point: 29-32° Boiling Point: 127-129°/19mm Beilstein Reference: 971015 UN No.: 1759 MDL No.: MFCD00000173

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 127…129 °C/19 mm
Melting Pt: 29…32 °C
Density: 1.881 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR