Cholic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
MDL: MFCD00003672
Beilstein Registry No.: 2822009
Optical Rotation: +36°C (c=0.6 in ethanol)
Purity: 98+%
CAS No.: 81-25-4
Molecular Formula: C24H40O5
Formula Weight: 408.58
Melting Point: 199-202°
Beilstein Reference: 2822009
Merck Reference: 14,2203
Optical Rotation: +35 ± 2° (c=0.6 in ethanol)
MDL No.: MFCD00003672
Formula:
C₂₄H₄₀O₅ MW: 408.58 g/mol Boiling Pt: 545 °C (1013 hPa) Melting Pt: 199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003672 CAS Number: 81-25-4 EINECS: 201-337-8 Merck Index: 13,02223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...