Undecanoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hendecanoic acid, Undecylic acid

A11244-30 A11244-18
AAA11244-30EA 114 USD
AAA11244-30 AAA11244-18
Undecanoic acid 98%
Undecanoic acid

An unsaturated fatty acid


Purity: 98% CAS No.: 112-37-8 Molecular Formula: CH3(CH2)9CO2H Formula Weight: 186.30 Flash Point: 110°(230°F) Melting Point: 26-29° Boiling Point: 283-285° Beilstein Reference: 1759287 MDL No.: MFCD00002730

Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 283…285 °C
Melting Pt: 26…29 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002730
CAS Number: 112-37-8
EINECS: 203-964-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR