3-Chloro-4-fluorobenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Chloro-4-fluorobenzene-1-sulphonyl chloride

A11222-06 A11222-14
AAAA11222-22EA 904.02 USD
AAAA11222-22 AAA11222-06 AAA11222-14
3-Chloro-4-fluorobenzenesulfonyl chloride 98%
3-Chloro-4-fluorobenzenesulphonyl chloride

MDL: MFCD00041444


Purity: 98% CAS No.: 91170-93-3 Molecular Formula: C6H3Cl2FO2S Formula Weight: 229.06 Flash Point: 110°(230°F) Refractive Index: 1.5520 Boiling Point: 104-105°/1mm UN No.: 3265 MDL No.: MFCD00041444

Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
Boiling Pt: 104…105 °C/1 mm
Density: 1.61 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041444
CAS Number: 91170-93-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR