2-Formylbenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11201-0B A11201-18 A11201-30
AAA11201-0BEA 780 USD
AAA11201-0B AAA11201-18 AAA11201-30
2-Formylbenzoic acid 98+%
2-Formylbenzoic acid

MDL: MFCD00003336 Beilstein Registry No.: 742381


Purity: 98+% CAS No.: 119-67-5 Molecular Formula: C8H6O3 Formula Weight: 150.13 Flash Point: 198°(388°F) Melting Point: 97-101° Beilstein Reference: 742381 MDL No.: MFCD00003336

Formula: C₈H₆O₃
Boiling Pt: 101 °C (0,4 mmHg)
Melting Pt: 97…101 °C
Density: 1,404
Flash Pt: 198 °C(388 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003336
CAS Number: 119-67-5
EINECS: 204-342-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR