1-Hexadecanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00004760
Beilstein Registry No.: 1748475
Soluble in alcohol, chloroform, ether
Purity: 98%
CAS No.: 36653-82-4
Molecular Formula: CH3(CH2)15OH
Formula Weight: 242.45
Flash Point: 135°(275°F)
Physical Form: Crystalline
Density: 0.817
Melting Point: 47-51°
Boiling Point: 190°/15mm
Solubility: Soluble in alcohol, chloroform, ether
Beilstein Reference: 1748475
Merck Reference: 14,2028
MDL No.: MFCD00004760
Formula:
H₃C(CH₂)₁₄CH₂OH Boiling Pt: 190 °C/15 mm Melting Pt: 47…51 °C Density: 0.817 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004760 CAS Number: 36653-82-4 EINECS: 253-149-0 Merck Index: 14,2028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...