Glutaric anhydride, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
A11152-18
A11152-30
A11152-0B
A11152-22
AAA11152-18EA
68.55
USD
AAA11152-18
AAA11152-30
AAAA11152-0B
AAAA11152-22
Glutaric anhydride, anhydrous 98%
Glutaric anhydride
MDL: MFCD00006679
Beilstein Registry No.: 110051
Purity: 98%
CAS No.: 108-55-4
Molecular Formula: C5H6O3
Formula Weight: 114.10
Flash Point: 178°(352°F)
Physical Form: Crystals
Density: 1.411
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 52-57°
Boiling Point: 291°
Beilstein Reference: 110051
MDL No.: MFCD00006679
Formula:
C₅H₆O₃ MW: 114.1 g/mol Boiling Pt: 291 °C Melting Pt: 52…57 °C Density: 1.411 g/cm³ (20 °C) Flash Pt: 178 °C(352 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006679 CAS Number: 108-55-4 EINECS: 203-593-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...