4-Bromoveratrole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00008381
Beilstein Registry No.: 2046884
Purity: 97%
CAS No.: 2859-78-1
Molecular Formula: C8H9BrO2
Formula Weight: 217.06
Flash Point: 109°(228°F)
Refractive Index: 1.5730
Boiling Point: 255-256°
Beilstein Reference: 2046884
MDL No.: MFCD00008381
Formula:
C₈H₉BrO₂ Boiling Pt: 255…256 °C Density: 1.5068 g/cm³ (20 °C) Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008381 CAS Number: 2859-78-1 EINECS: 220-677-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...