1-Naphthylacetic acid 96% may cont. up to 4% 2-isomer

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Planofix, 1-Naphthaleneacetic acid

A11069-18 A11069-30 A11069-0B
AAA11069-18EA 32.7 USD
AAA11069-18 AAA11069-30 AAA11069-0B
1-Naphthylacetic acid 96% may cont. up to 4% 2-isomer
1-Naphthylacetic acid

MDL: MFCD00004046 Beilstein Registry No.: 1308415


Purity: 96% CAS No.: 86-87-3 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 126-133° Beilstein Reference: 1308415 Merck Reference: 14,6371 MDL No.: MFCD00004046

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 126…133 °C
Density: ~ 1.263 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004046
CAS Number: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR