Propionyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00000745
Beilstein Registry No.: 385632
Purity: 98%
CAS No.: 79-03-8
Molecular Formula: C3H5ClO
Formula Weight: 92.53
Flash Point: 11°(51°F)
Refractive Index: 1.4040
Odor: Unpleasant
Melting Point: -94°
Boiling Point: 77-79°
Beilstein Reference: 385632
Merck Reference: 14,7828
UN No.: 1815
MDL No.: MFCD00000745
Formula:
C₃H₅ClO Boiling Pt: 77…79 °C Melting Pt: –94 °C Density: 1.06 g/cm³ (20 °C) Flash Pt: 11 °C (51 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000745 CAS Number: 79-03-8 EINECS: 201-170-0 UN: 1815 ADR: 3,II Merck Index: 12,08013 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...