3-Chlorobenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10876-36
A10876-14
A10876-22
AAAA10876-36EA
229
USD
AAAA10876-36
AAA10876-14
AAA10876-22
3-Chlorobenzoic acid 99%
3-Chlorobenzoic acid
MDL: MFCD00002491
Beilstein Registry No.: 907218
Purity: 99%
CAS No.: 535-80-8
Molecular Formula: C7H5ClO2
Formula Weight: 156.57
Odor: Odorless
Melting Point: 154-160°
Boiling Point: 274-276°
Beilstein Reference: 907218
Merck Reference: 14,2123
MDL No.: MFCD00002491
Formula:
C₇H₅ClO₂ MW: 156.57 g/mol Boiling Pt: 274…276 °C Melting Pt: 154…160 °C Density: 1.496 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002491 CAS Number: 535-80-8 EINECS: 208-618-4 Merck Index: 13,02142 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...