1-Methylpiperazine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10837-0F A10837-AE A10837-AP
AAA10837-0FEA 490 USD
AAA10837-0F AAA10837-AE AAA10837-AP
1-Methylpiperazine 98+%
1-Methylpiperazine

MDL: MFCD00005966 Beilstein Registry No.: 102724


Purity: 98+% CAS No.: 109-01-3 Molecular Formula: C5H12N2 Formula Weight: 100.17 Flash Point: 34°(93°F) Refractive Index: 1.4655 Odor: Amine-like Melting Point: -6° Boiling Point: 137-139° Beilstein Reference: 102724 UN No.: 2734 MDL No.: MFCD00005966

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 137…139 °C
Melting Pt: –6 °C
Density: 0.9085 g/cm³ (20 °C)
Flash Pt: 34 °C (93 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR