Bromotrichloromethane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10824-22
A10824-36
AAA10824-22EA
51.43
USD
AAA10824-22
AAA10824-36
Bromotrichloromethane 97%
Bromotrichloromethane
MDL: MFCD00000783
Beilstein Registry No.: 1732543
Fieser: 1,80 2,42 7,40 20,59
Purity: 97%
CAS No.: 75-62-7
Molecular Formula: CBrCl3
Formula Weight: 198.28
Flash Point: None
Refractive Index: 1.5060
Odor: Chloroform-like
Melting Point: -6°
Boiling Point: 104-105°
Beilstein Reference: 1732543
UN No.: 2810
MDL No.: MFCD00000783
Formula:
BrCl₃C MW: 198.27 g/mol Boiling Pt: 104…105 °C Melting Pt: –6 °C Density: 2.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000783 CAS Number: 75-62-7 EINECS: 200-886-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...