Anthranilohydrazide 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Aminobenzoic hydrazide, 2-Aminobenzoic acid hydrazide, 2-Aminobenzohydrazide, 2-Aminobenzoylhydrazide, Anthraniloyl Hydrazine, 2-Aminobenzene-1-carbohydrazide, 2-Aminobenzhydrazide

A10805-18 A10805-09
AAA10805-18EA 161 USD
AAA10805-18 AAA10805-30 AAA10805-09
Anthranilohydrazide 98+%
Anthranilohydrazide

MDL: MFCD00017056 Beilstein Registry No.: 742811


Purity: 98+% CAS No.: 1904-58-1 Molecular Formula: C7H9N3O Formula Weight: 151.17 Melting Point: 122-124° Beilstein Reference: 742811

Formula: C₇H₉N₃O
MW: 151.17 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00017056
CAS Number: 1904-58-1
EINECS: 217-604-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR