Pivalic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Dimethylpropionic acid, Trimethylacetic acid
A10776-0C
A10776-0E
A10776-36
AAA10776-0CEA
691
USD
AAA10776-0C
AAA10776-0E
AAA10776-36
Pivalic acid 99%
Pivalic acid
MDL: MFCD00004194
Beilstein Registry No.: 969480
Soluble in alcohol, ether. Slightly soluble in water
Purity: 99%
CAS No.: 75-98-9
Molecular Formula: (CH3)3CCO2H
Formula Weight: 102.13
Flash Point: 63°(145°F)
Odor: Pungent
Boiling Point: 163-165°
Solubility: Soluble in alcohol, ether. Slightly soluble in water
Beilstein Reference: 969480
Merck Reference: 14,7511
UN No.: 3261
MDL No.: MFCD00004194
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 163…165 °C Melting Pt: 31…35 °C Density: 0.91 g/cm³ (20 °C) Flash Pt: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004194 CAS Number: 75-98-9 EINECS: 200-922-5 UN: 3261 ADR: 8,III Merck Index: 12,07667 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...