ω-Chloroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloroacetophenone, α-Chloroacetophenone, Phenacyl chloride

A10749-36 A10749-22
AAA10749-36EA 276 USD
AAA10749-36 AAA10749-22
ω-Chloroacetophenone 98%
ω-Chloroacetophenone

MDL: MFCD00000933 Beilstein Registry No.: 507950


Purity: 98% CAS No.: 532-27-4 Molecular Formula: C8H7ClO Formula Weight: 154.60 Flash Point: 118°(244°F) Odor: Irritating Boiling Point: 243-244° Beilstein Reference: 507950 Merck Reference: 14,2115 UN No.: 1697 MDL No.: MFCD00000933

Formula: (C₆H₅)C(O)CH₂Cl
MW: 154.6 g/mol
Boiling Pt: 243…244 °C
Melting Pt: 52…56 °C
Density: 1.324 g/cm³ (15 °C)
Flash Pt: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000933
CAS Number: 532-27-4
EINECS: 208-531-1
UN: 1697
ADR: 6.1,II
Merck Index: 13,02132

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR