Benzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

A10739-0B A10739-30 A10739-0I
AAA10739-0BEA 93.69 USD
AAA10739-0B AAA10739-30 AAA10739-0I
Benzophenone 99%
Benzophenone

MDL: MFCD00003076 Beilstein Registry No.: 1238185


Purity: 99% CAS No.: 119-61-9 Molecular Formula: C13H10O Formula Weight: 182.22 Flash Point: 138°(280°F) Physical Form: Cryst flakes or powder Density: 1.111 Appearance: White Odor: Pleasant Melting Point: 47-50° Boiling Point: 305° Beilstein Reference: 1238185 Merck Reference: 14,1098 UN No.: 3077 MDL No.: MFCD00003076

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47 - 49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR