ω-Bromo-3-methoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00000199
Beilstein Registry No.: 510128
Purity: 98%
CAS No.: 5000-65-7
Molecular Formula: C9H9BrO2
Formula Weight: 229.08
Storage Temperature: 2°C to 8°C
Odor: Irritating
Beilstein Reference: 510128
UN No.: 3261
MDL No.: MFCD00000199
Formula:
C₉H₉BrO₂ Boiling Pt: 285 °C (1013 hPa) Melting Pt: 63 - 64.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000199 CAS Number: 5000-65-7 EINECS: 225-666-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...