4'-Isobutylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Isobutylacetophenone

A10624-18 A10624-30
AAA10624-18EA 150.22 USD
AAA10624-18 AAA10624-30
4'-Isobutylacetophenone 97%
4'-Isobutylacetophenone

MDL: MFCD00027393 Beilstein Registry No.: 1935275


Purity: 97% CAS No.: 38861-78-8 Molecular Formula: C12H16O Formula Weight: 176.27 Flash Point: 54°(129°F) Refractive Index: 1.5180 Beilstein Reference: 1935275 UN No.: 1224 MDL No.: MFCD00027393

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Density: 0.952
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027393
CAS Number: 38861-78-8
EINECS: 254-159-8
UN: 1224
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR