1,3-Diiodopropane 98+% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylene diiodide

Stabilization: Stabilised with copper

A10616-30 A10616-18
AAA10616-30EA 295 USD
AAA10616-30 AAA10616-18
1,3-Diiodopropane 98+% stabilized
1,3-Diiodopropane

MDL: MFCD00001097 Beilstein Registry No.: 1731117


Purity: 98+% CAS No.: 627-31-6 Molecular Formula: I(CH2)3I Formula Weight: 295.89 Flash Point: 110°(230°F) Refractive Index: 1.6415 Melting Point: -20° Boiling Point: 224° Beilstein Reference: 1731117 MDL No.: MFCD00001097

Formula: C₃H₆I₂
MW: 295.89 g/mol
Boiling Pt: 224 °C
Melting Pt: –20 °C
Density: 2.59617 g/cm³ (4 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001097
CAS Number: 627-31-6
EINECS: 210-993-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR