5-Methylbenzotriazole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00005702
Beilstein Registry No.: 116658
Purity: 98+%
CAS No.: 136-85-6
Molecular Formula: C7H7N3
Formula Weight: 133.15
Odor: Odorless
pH: 5.3
Melting Point: 77-84°
Boiling Point: 210-212°/12mm
Beilstein Reference: 116658
MDL No.: MFCD00005702
Formula:
C₇H₇N₃ Boiling Pt: 210…212 °C/12 mm Melting Pt: 77…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005702 CAS Number: 136-85-6 EINECS: 205-265-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...