Nitrobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10585-0C
A10585-0E
A10585-36
AAA10585-0CEA
317
USD
AAA10585-0C
AAA10585-0E
AAA10585-36
Nitrobenzene 99%
Nitrobenzene
MDL: MFCD00007043
Beilstein Registry No.: 507540
Fieser: 2,295 7,251 8,358 19,229
Purity: 99%
CAS No.: 98-95-3
Molecular Formula: C6H5NO2
Formula Weight: 123.11
Flash Point: 87°(188°F)
Refractive Index: 1.5520
Melting Point: 5-6°
Boiling Point: 210-212°
Beilstein Reference: 507540
Merck Reference: 14,6588
UN No.: 1662
MDL No.: MFCD00007043
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Boiling Pt: 210…212 °C Melting Pt: 5…6 °C Density: 1.2037 g/cm³ (20 °C) Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007043 CAS Number: 98-95-3 EINECS: 202-716-0 UN: 1662 ADR: 6.1,II Merck Index: 13,06621 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...