4-Bromothiophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

A10555-30 A10555-18 A10555-09
AAAA10555-30EA 777 USD
AAAA10555-30 AAA10555-18 AAA10555-09
4-Bromothiophenol 98%
4-Bromothiophenol

MDL: MFCD00004845 Beilstein Registry No.: 507031


Purity: 98% CAS No.: 106-53-6 Molecular Formula: C6H5BrS Formula Weight: 189.08 Odor: Unpleasant Melting Point: 72-78° Boiling Point: 239° Beilstein Reference: 507031 UN No.: 2923 MDL No.: MFCD00004845

Formula: C₆H₅BrS
Boiling Pt: 239 °C
Melting Pt: 72…78 °C
Density: 1.526 g/cm³ (83 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004845
CAS Number: 106-53-6
EINECS: 203-407-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR