1-Bromopropane 99% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Solvent 38, n-Propyl bromide
A10461-0J
A10461-AK
A10461-AU
AAA10461-0JEA
520
USD
AAA10461-0J
AAA10461-AK
AAA10461-AU
1-Bromopropane 99% ACS
1-Bromopropane
MDL: MFCD00000254
Beilstein Registry No.: 505936
Purity: 99%
CAS No.: 106-94-5
Molecular Formula: C3H7Br
Formula Weight: 123.00
Flash Point: 21°(70°F)
Refractive Index: 1.4340
Melting Point: -110°
Boiling Point: 70-71°
Beilstein Reference: 505936
Merck Reference: 14,7845
UN No.: 2344
MDL No.: MFCD00000254
Formula:
BrH₂CCH₂CH₃ MW: 122.99 g/mol Boiling Pt: 70…71 °C Melting Pt: –110 °C Density: 1.354 g/cm³ (20 °C) Flash Pt: 21 °C (70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000254 CAS Number: 106-94-5 EINECS: 203-445-0 UN: 2344 ADR: 3,III Merck Index: 12,08030 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...