Dibromomethane 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methylenbromide
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 50 ppm
A10456-36
A10456-0E
A10456-22
AAA10456-36EA
47
USD
AAA10456-36
AAA10456-0E
AAA10456-22
Dibromomethane 99% stabilized
Dibromomethane
MDL: MFCD00000168
Beilstein Registry No.: 969143
Fieser: 11,157 13,93 14,112 16,107 18,122
Purity: 99%
CAS No.: 74-95-3
Molecular Formula: CH2Br2
Formula Weight: 173.85
Refractive Index: 1.5410
Melting Point: -53°
Boiling Point: 96-98°
Beilstein Reference: 969143
Merck Reference: 14,6061
UN No.: 2664
MDL No.: MFCD00000168
Formula:
Br₂CH₂ MW: 173.83 g/mol Boiling Pt: 96…98 °C Melting Pt: –53 °C Density: 2.497 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000168 CAS Number: 74-95-3 EINECS: 200-824-2 UN: 2664 ADR: 6.1,III Merck Index: 12,06138 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...