2-Fluoro-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00039215
Beilstein Registry No.: 1857413
Purity: 98%
CAS No.: 443-88-9
Molecular Formula: C8H9F
Formula Weight: 124.16
Flash Point: 30°(86°F)
Refractive Index: 1.4790
Odor: Aromatic
Boiling Point: 147-148°
Beilstein Reference: 1857413
UN No.: 1993
MDL No.: MFCD00039215
Formula:
C₈H₉F Boiling Pt: 165 - 167 °C (1013 hPa) Density: 0.988 g/cm³ (20 °C) Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039215 CAS Number: 443-88-9 EINECS: 207-144-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...