4-Nitroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00007858
Beilstein Registry No.: 508690
Soluble in hot water and alcohol
Purity: 98%
CAS No.: 100-01-6
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Flash Point: 165°(329°F)
Odor: Amine-like
Boiling Point: 260°/100mm
Solubility: Soluble in hot water and alcohol
Beilstein Reference: 508690
Merck Reference: 14,6584
UN No.: 1661
MDL No.: MFCD00007858
Formula:
C₆H₆N₂O₂ Boiling Pt: 260 °C/100 mm Melting Pt: 147…149 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 165 °C (329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007858 CAS Number: 100-01-6 EINECS: 202-810-1 UN: 1661 ADR: 6.1,II Merck Index: 12,06681 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...