Benzyltributylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Benzyl-N,N-dibutylbutan-1-aminium chloride

A10345-0E A10345-36
AAA10345-0EEA 384 USD
AAA10345-0E AAA10345-36
Benzyltributylammonium chloride 98%
Benzyltributylammonium chloride

MDL: MFCD00011849 Beilstein Registry No.: 3776210 Fieser: 6,41


Purity: 98% CAS No.: 23616-79-7 Molecular Formula: [CH3(CH2)3]3N(CH2C6H5)Cl Formula Weight: 311.94 Melting Point: 156-162° Beilstein Reference: 3776210

Formula: C₁₉H₃₄ClN
MW: 311.94 g/mol
Melting Pt: 156…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011849
CAS Number: 23616-79-7
EINECS: 245-787-3
UN: UN3263

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR