N-Chlorosuccinimide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Chloropyrrolidine-2,5-dione
A10310-0B
A10310-18
A10310-30
AAA10310-0BEA
163
USD
AAA10310-0B
AAA10310-18
AAA10310-30
N-Chlorosuccinimide 98%
N-Chlorosuccinimide
Regioselective chlorination and oxidizing reagent
Purity: 98%
CAS No.: 128-09-6
Molecular Formula: C4H4ClNO2
Formula Weight: 133.53
Physical Form: Powder
Density: 1.65
Appearance: White
Odor: Chlorine-like
Melting Point: 146-150°
Beilstein Reference: 113915
Merck Reference: 14,2164
UN No.: 3261
MDL No.: MFCD00005511
Regioselective chlorination and oxidizing reagent
Formula:
C₄H₄ClNO₂ MW: 133.53 g/mol Boiling Pt: 216.5 °C (1013 hPa) Melting Pt: 146…150 °C Density: 1.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005511 CAS Number: 128-09-6 EINECS: 204-878-8 UN: 3261 ADR: 8,III Merck Index: 13,02183 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...