1,4-Dinitrobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10293-14
A10293-22
A10293-06
AAA10293-14EA
184
USD
AAA10293-14
AAA10293-22
AAA10293-06
1,4-Dinitrobenzene 98+%
1,4-Dinitrobenzene
MDL: MFCD00007314
Beilstein Registry No.: 1105828
Purity: 98+%
CAS No.: 100-25-4
Molecular Formula: C6H4N2O4
Formula Weight: 168.11
Flash Point: 150°(302°F)
Melting Point: 172-176°
Boiling Point: 299°
Beilstein Reference: 1105828
Merck Reference: 14,3273
UN No.: 3443
MDL No.: MFCD00007314
Formula:
C₆H₄N₂O₄ MW: 168.11 g/mol Boiling Pt: 299 Melting Pt: 172 Density: 1.625 Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007314 CAS Number: 100-25-4 EINECS: 202-833-7 UN: 3443 ADR: 6.1 II Merck Index: 14,3273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...