4-Bromo-2-fluoroaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00010221
Beilstein Registry No.: 2081089
Purity: 98+%
CAS No.: 367-24-8
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Flash Point: 104°(219°F)
Melting Point: 38-42°
Boiling Point: 103-108°/13mm
Beilstein Reference: 2081089
UN No.: 2811
MDL No.: MFCD00010221
Formula:
C₆H₅BrFN Boiling Pt: 75 - 80 °C (0.3 -rr) Melting Pt: 41 - 42 °C Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010221 CAS Number: 367-24-8 EINECS: 206-685-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...