4-Amino-3-hydroxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10207-06
A10207-22
A10207-14
AAA10207-06EA
42
USD
AAA10207-06
AAAA10207-22
AAA10207-14
4-Amino-3-hydroxybenzoic acid 98%
4-Amino-3-hydroxybenzoic acid
MDL: MFCD00017094
Beilstein Registry No.: 509859
Purity: 98%
CAS No.: 2374-03-0
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 215° dec.
Beilstein Reference: 509859
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Melting Pt: ca 215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00017094 CAS Number: 2374-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...