2-Heptanone 99%

Supplier: Thermo Scientific Chemicals
Warning

A10200-AE A10200-AP
AAA10200-AEEA 41 USD
AAA10200-AE AAA10200-AP
2-Heptanone 99%
2-Heptanone

MDL: MFCD00009513 Beilstein Registry No.: 1699063


Purity: 99% CAS No.: 110-43-0 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 47°(116°F) Refractive Index: 1.4090 Odor: Unpleasant Melting Point: -35° Boiling Point: 149-150° Beilstein Reference: 1699063 Merck Reference: 14,4663 UN No.: 1110 MDL No.: MFCD00009513

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 149…150 °C
Melting Pt: –35 °C
Density: 0.8111 g/cm³ (20 °C)
Flash Pt: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009513
CAS Number: 110-43-0
EINECS: 203-767-1
UN: 1110
ADR: 3,III
Merck Index: 13,04683

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR