(4-Ethoxyphenyl)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10129-09 A10129-18 A10129-14
AAA10129-09EA 44.7 USD
AAA10129-09 AAA10129-18 AAAA10129-14
(4-Ethoxyphenyl)acetic acid 98+%
(4-Ethoxyphenyl)acetic acid

MDL: MFCD00004346 Beilstein Registry No.: 1103143


Purity: 98+% CAS No.: 4919-33-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 87-91° Beilstein Reference: 1103143

Formula: (C₆H₄(OCH₂CH₃))CH₂COOH
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00004346
CAS Number: 4919-33-9
EINECS: 225-545-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR