Toluene-α-sulfonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

A10124-06 A10124-22 A10124-14
AAA10124-06EA 40.6 USD
AAA10124-06 AAA10124-22 AAA10124-14
Toluene-α-sulfonyl chloride 99%
Toluene-α-sulphonyl chloride

MDL: MFCD00007455 Beilstein Registry No.: 972806


Purity: 99% CAS No.: 1939-99-7 Molecular Formula: C6H5CH2SO2Cl Formula Weight: 190.65 Melting Point: 89-94° Boiling Point: 120°/10mm UN No.: 3096 MDL No.: MFCD00007455

Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 120 °C (10 mmHg)
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007455
CAS Number: 1939-99-7
EINECS: 217-717-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR