Strontium chromate ≥95%

Supplier: Thermo Scientific Chemicals
Danger

089026-30 089026-A1
AA89026-30EA 74.2 USD
AA89026-30 AA89026-A1
Strontium chromate ≥95%
Strontium chromate

MDL: MFCD00049549 Soluble in boiling water, dilute HCl2 dilute CH3CO2H


CAS No.: 7789-06-2 Molecular Formula: SrCrO4 Formula Weight: 203.61 Physical Form: Powder Density: 3.895 Odor: Odorless Merck Reference: 14,8841 UN No.: 3087 MDL No.: MFCD00049549

Formula: CrO₄Sr
Density: 3.895
Storage Temperature: Ambient
MDL Number: MFCD00049549
CAS Number: 7789-06-2
EINECS: 232-142-6
UN: 3087
ADR: 5.1,III
Merck Index: 14,8841

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR