Yttrium(III) fluoride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Yttrium trifluoride, Yttrium(III) fluoride
083106-14
083106-22
AA83106-14EA
60.4
USD
AA83106-14
AA83106-22
Yttrium(III) fluoride 98%
Yttrium(III) fluoride
MDL: MFCD00049714
Purity: 98%
CAS No.: 13709-49-4
Molecular Formula: F3Y
Formula Weight: 145.90
Physical Form: -325 Mesh Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 1387°
UN No.: 3288
MDL No.: MFCD00049714
Formula:
F₃Y MW: 145.9 Boiling Pt: 2227 Melting Pt: 1387 Density: 4.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049714 CAS Number: 13709-49-4 EINECS: 237-257-5 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...