Trimethyltin hydroxide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00013929
Notes: Special handling precautions required. View MSDS prior to purchase. Begins to sublime at 80°C
Purity: 98%
CAS No.: 56-24-6
Molecular Formula: C3H10OSn
Formula Weight: 180.80
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Pungent
Melting Point: 114-118°
UN No.: 3146
MDL No.: MFCD00013929
Formula:
C₃H₁₀OSn Melting Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013929 CAS Number: 56-24-6 UN: 3146 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...