Trimethyltin chloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorotrimethylstannane
071166-04
071166-18
071166-09
AA71166-04EA
35
USD
AA71166-04
AA71166-18
AA71166-09
Trimethyltin chloride
Trimethyltin chloride
MDL: MFCD00000520
Notes: Special handling precautions required. View MSDS prior to purchase.
CAS No.: 1066-45-1
Molecular Formula: (CH3)3SnCl
Formula Weight: 199.25
Flash Point: 97°(207°F)
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Unpleasant
Melting Point: 37-39°
Boiling Point: 154°
UN No.: 3146
MDL No.: MFCD00000520
Formula:
C₃H₉ClSn MW: 199.27 g/mol Boiling Pt: 154 °C Melting Pt: 37…39 °C Flash Pt: 97 °C (207 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000520 CAS Number: 1066-45-1 EINECS: 213-917-8 UN: 3146 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...