Zirconium ≥99.2% (metals basis excluding Hf), plate 100x100 mm (3.9x3.9 in), Thickness: 6.4 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

044261-KF 044261-KJ
AA44261-KFEA 764 USD
AA44261-KF AA44261-KJ
Zirconium ≥99.2% (metals basis excluding Hf), plate 100x100 mm (3.9x3.9 in), Thickness: 6.4 mm (0.25 in)
Zirconium

MDL: MFCD00011303


Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈416g/100x100mm, Temper: annealed Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR