1-Aminopyrene ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyren-1-ylamine, Pyren-1-amine

043968-02 043968-04
AA43968-02EA 54.5 USD
AA43968-02 AA43968-04
1-Aminopyrene ≥97%
1-Aminopyrene

MDL: MFCD00004140


Purity: 97% CAS No.: 1606-67-3 Molecular Formula: C16H11N Formula Weight: 217.27 Physical Form: Powder Melting Point: 116-119° MDL No.: MFCD00004140

Formula: C₁₆H₁₁N
MW: 217.27 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004140
CAS Number: 1606-67-3
EINECS: 216-521-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR