Silicon carbide, β-phase ≥95%, nanopowder

Supplier: Thermo Scientific Chemicals
Warning

043884-06 043884-14 043884-22
AA43884-06EA 82.5 USD
AA43884-06 AA43884-14 AA43884-22
Silicon carbide, β-phase ≥95%, nanopowder
Silicon carbide, β-phase

MDL: MFCD00049531 Insoluble in water, alcohol. Soluble in fused alkali


Purity: 95% min CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 30nm APS Powder, S.A. 80m2/g Appearance: Grey to black Odor: Odorless Melting Point: 2700° Solubility: Insoluble in water, alcohol. Soluble in fused alkali Merck Reference: 14,8492 MDL No.: MFCD00049531

Formula: SiC
MW: 40.10 g/mol
Melting Pt: 2700 °C
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR