Diarsenic triselenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Arsenic(III) selenide

043748-06 043748-14
AA43748-06EA 153 USD
AA43748-06 AA43748-14
Diarsenic triselenide ≥99.999% (metals basis)
Diarsenic triselenide

MDL: MFCD00014166


Purity: 99.999% CAS No.: 1303-36-2 Molecular Formula: As2Se3 Formula Weight: 386.72 Physical Form: -160 Mesh Powder Density: 4.75 Melting Point: 260° Merck Reference: 14,805 UN No.: 3077 MDL No.: MFCD00014166

Formula: As₂Se₃
MW: 386.72 g/mol
Melting Pt: 382.35 - 383.35 °C
Density: 4.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014166
CAS Number: 1303-36-2
EINECS: 215-119-5
UN: 3077
ADR: 9,III
Merck Index: 12,00845

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR