Copper(I) iodide ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cuprous iodide
043153-14
043153-06
043153-22
AA43153-14EA
299
USD
AA43153-14
AA43153-06
AA43153-22
Copper(I) iodide ≥99.998% (metals basis), Puratronic®
Copper(I) iodide
MDL: MFCD00010978
Purity: 99.998%
CAS No.: 7681-65-4
Molecular Formula: CuI
Formula Weight: 190.45
Physical Form: Chunks/powder
Density: 5.63
Refractive Index: 2.346
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Pale brown
Odor: Weak
Melting Point: 605°
Boiling Point: ca 1290°
Merck Reference: 14,2662
UN No.: 3077
MDL No.: MFCD00010978
Formula:
CuI MW: 190.45 g/mol Boiling Pt: ∼1290 °C Melting Pt: 605 °C Density: 5.62 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9,III Merck Index: 13,02692 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...