Lanthanum(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lanthanum trichloride hydrate
043120-22
043120-36
AA43120-22EA
84.5
USD
AA43120-22
AA43120-36
Lanthanum(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) chloride hydrate
MDL: MFCD00149753
Purity: 99.9%
CAS No.: 20211-76-1
Molecular Formula: LaCl3.xH2O
Formula Weight: 245.27(anhy)
Physical Form: Crystalline
Odor: Odorless
pH: 5
Melting Point: 91° dec.
Merck Reference: 14,5363
MDL No.: MFCD00149753
Formula:
Cl₃La·H₂O MW: 263.28 g/mol Boiling Pt: 1000 °C (1013 hPa) Melting Pt: 91 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149753 CAS Number: 20211-76-1 EINECS: 233-237-5 Merck Index: 14,5363 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...