Cesium acetate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid cesium salt

043095-18 043095-30
AA43095-18EA 87.5 USD
AA43095-18 AA43095-30
Cesium acetate ≥99% (metals basis)
Cesium acetate

MDL: MFCD00013056


Purity: 99% CAS No.: 3396-11-0 Molecular Formula: CsOOCCH3 Formula Weight: 191.95 Physical Form: Crystalline Powder Odor: Weak MDL No.: MFCD00013056

Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194
Storage Temperature: Ambient
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR