Germanium(IV) ethoxide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Germanium tetraethanolate, Germanic acid tetraethyl ester, Tetraethyl orthogermanate, Germanium tetraethoxide, Germanium (IV) ethanolate
Preparation of GeO2 by CVD; intermediate in sol-gel synthesis
Purity: 99.995%
CAS No.: 14165-55-0
Molecular Formula: C8H20GeO4
Formula Weight: 252.84
Flash Point: 49°(121°F)
Physical Form: Liquid
Density: 1.14
Refractive Index: 1.4070
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Alcohol-like
Melting Point: -72°
Boiling Point: 185.5°
UN No.: 1993
MDL No.: MFCD00015121
Preparation of GeO2 by CVD; intermediate in sol-gel synthesis
Formula:
C₈H₂₀GeO₄ MW: 252.85 g/mol Boiling Pt: 185.5 °C Melting Pt: –72 °C Density: 1.14 g/cm³ (20 °C) Flash Pt: 49 °C (121 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015121 CAS Number: 14165-55-0 EINECS: 238-007-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...