Platinum rhodium alloy (95.2:4.8 w%), foil, Thickness 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

042686-FF 042686-FI
AA42686-FFEA 218 USD
AA42686-FF AA42686-FI
Platinum rhodium alloy (95.2:4.8 w%), foil, Thickness 0.05 mm (0.002 in)
Platinum rhodium alloy (95.2:4.8 w%)

MDL: MFCD00798552


Molecular Formula: Pt:Ru; 95.2:4.8 wt% Physical Form: ≈0.66g/25x25mm Odor: Odorless MDL No.: MFCD00798552

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR